accessibility menu, dialog, popup

UserName

ClBr

CAS RN 179526-95-5

IUPAC Name: 2-bromo-4'-chloro-1,1'-biphenyl
Synonyms: 2-bromo-4'-chloro-1,1'-biphenyl
Molecular Weight (g/mol): 267.55
Molecular Formula: C12H8BrCl
InChi Key: WSHZWUXRWQVZQP-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CC=CC=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results