missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chlorobenzoic acid, 97%

Catalog No. p-7053382
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5977703 1 g
AAH5977706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5977703 Supplier Thermo Scientific Chemicals Supplier No. H5977703

CAS: 936-08-3 | C7H4BrClO2 | 235.461 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 936-08-3
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00672930
InChI Key USMQLFCVCDEXAK-UHFFFAOYSA-N
Synonym 2-bromo-4-chloro-benzoic acid, benzoic acid, 2-bromo-4-chloro, 2-bromo-4-chlorobenzoicacid, pubchem4028, acmc-209rmm, p-chloro bromo benzoic acid, ksc486m1d, benzoicacid, 2-bromo-4-chloro, 2-bromo-4-chlorobenzoic acid
PubChem CID 33126
IUPAC Name 2-bromo-4-chlorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)Br)C(=O)O

Specifications

Melting Point 157°C to 161°C
Quantity 1 g
UN Number UN2811
Formula Weight 235.46
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Bromo-4-chlorobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only