accessibility menu, dialog, popup

UserName

ClBrOHO

CAS RN 936-08-3

IUPAC Name: 2-bromo-4-chlorobenzoic acid
Synonyms: 2-bromo-4-chlorobenzoic acid
Molecular Weight (g/mol): 235.46
Molecular Formula: C7H4BrClO2
InChi Key: USMQLFCVCDEXAK-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Br)C=C(Cl)C=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)