missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-chloroaniline 98.0+%, TCI America™
Supplier: TCI America B226825G
Specifications
| 2-Bromo-4-chloroaniline | |
| 66°C | |
| 127°C | |
| 25 g | |
| SYTBIFURTZACKR-UHFFFAOYSA-N | |
| 2-bromo-4-chloroaniline | |
| 70110 | |
| ≥98.0% (GC) |
| 873-38-1 | |
| White-Yellow | |
| C6H5BrClN | |
| benzenamine, 2-bromo-4-chloro, 1-amino-2-bromo-4-chlorobenzene, 2-bromo-4-chloro-phenylamine, 2-bromo-4-chloro aniline, 2-bromo-4-chlorophenylamine, pubchem3576, acmc-1bkdm, 2-bromo-4-chloro-aniline, 4-chloro-2-bromoaniline, 2-bromo-4-chloroaniline | |
| C1=CC(=C(C=C1Cl)Br)N | |
| 206.467 | |
| 206.47 | |
| Crystalline Powder |
Chemical Identifiers
| 873-38-1 | |
| 206.467 | |
| benzenamine, 2-bromo-4-chloro, 1-amino-2-bromo-4-chlorobenzene, 2-bromo-4-chloro-phenylamine, 2-bromo-4-chloro aniline, 2-bromo-4-chlorophenylamine, pubchem3576, acmc-1bkdm, 2-bromo-4-chloro-aniline, 4-chloro-2-bromoaniline, 2-bromo-4-chloroaniline | |
| 2-bromo-4-chloroaniline |
| C6H5BrClN | |
| SYTBIFURTZACKR-UHFFFAOYSA-N | |
| 70110 | |
| C1=CC(=C(C=C1Cl)Br)N |
Safety and Handling
TSCA : No