missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloroaniline 98.0+%, TCI America™

Catalog No. p-7119589
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22685G 5 g
B226825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22685G Supplier TCI America Supplier No. B22685G
Only null left

Chemical Identifiers

CAS 873-38-1
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
InChI Key SYTBIFURTZACKR-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-4-chloro, 1-amino-2-bromo-4-chlorobenzene, 2-bromo-4-chloro-phenylamine, 2-bromo-4-chloro aniline, 2-bromo-4-chlorophenylamine, pubchem3576, acmc-1bkdm, 2-bromo-4-chloro-aniline, 4-chloro-2-bromoaniline, 2-bromo-4-chloroaniline
PubChem CID 70110
IUPAC Name 2-bromo-4-chloroaniline
SMILES C1=CC(=C(C=C1Cl)Br)N

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 127°C
Quantity 5 g
Formula Weight 206.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-chloroaniline