missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-chloroacetophenone, 98%

CAS: 536-38-9 | C8H6BrClO | 233.489 g/mol

Supplier:  Thermo Scientific Chemicals A1260736

 View more versions of this product

Catalog No. AAA1260736


Only null left
Explore available promotions

Description

Description

2-Bromo-4'-chloroacetophenone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-chloroacetophenone is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications

Specifications

2-Bromo-4'-chloroacetophenone
94°C to 98°C
MFCD00000625
UN3261
14,2153
Insoluble in water.
C1=CC(=CC=C1C(=O)CBr)Cl
233.489
233.5
536-38-9
C8H6BrClO
500 g
607603
2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone
FLAYZKKEOIAALB-UHFFFAOYSA-N
2-bromo-1-(4-chlorophenyl)ethanone
68303
98%

Chemical Identifiers

536-38-9
233.489
FLAYZKKEOIAALB-UHFFFAOYSA-N
68303
C1=CC(=CC=C1C(=O)CBr)Cl
C8H6BrClO
MFCD00000625
2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone
2-bromo-1-(4-chlorophenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 208-631-5

RTECSNumber : AM5978800

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only