missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-chloroacetophenone, 98%

Catalog No. p-7020596
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1260714 25 g
AAA1260722 100 g
AAA1260736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1260714 Supplier Thermo Scientific Chemicals Supplier No. A1260714
Only null left

CAS: 536-38-9 | C8H6BrClO | 233.489 g/mol

2-Bromo-4'-chloroacetophenone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-chloroacetophenone is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 536-38-9
Molecular Formula C8H6BrClO
Molecular Weight (g/mol) 233.489
MDL Number MFCD00000625
InChI Key FLAYZKKEOIAALB-UHFFFAOYSA-N
Synonym 2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone
PubChem CID 68303
IUPAC Name 2-bromo-1-(4-chlorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)Cl

Specifications

Melting Point 94°C to 98°C
Quantity 25 g
UN Number UN3261
Beilstein 607603
Merck Index 14,2153
Solubility Information Insoluble in water.
Formula Weight 233.5
Percent Purity 98%
Chemical Name or Material 2-Bromo-4'-chloroacetophenone
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 208-631-5
RTECSNumber AM5978800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only