missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-dichloroaniline, 98+%

Catalog No. AAA1843614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1843614 25 g
AAA1843622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1843614 Supplier Thermo Scientific Chemicals Supplier No. A1843614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 697-86-9 | C6H4BrCl2N | 240.909 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 697-86-9
Molecular Formula C6H4BrCl2N
Molecular Weight (g/mol) 240.909
MDL Number MFCD00040936
InChI Key DTPADCOGQUOGHT-UHFFFAOYSA-N
Synonym pubchem3605, acmc-209ob3, 6-bromo-2,4-dichloroaniline, 2,4-dichloro-6-bromoaniline, 2-bromo-4,6-dichloro-aniline, 6-bromo-2,4-dichlorophenylamine, benzenamine,2-bromo-4,6-dichloro, 2-bromo-4,6-dichloroaniline, 2-bromanyl-4,6-bis chloranyl aniline
PubChem CID 2756901
IUPAC Name 2-bromo-4,6-dichloroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Br)Cl

Specifications

Melting Point 78°C to 82°C
Quantity 25 g
Beilstein 2089686
Formula Weight 240.92
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-4,6-dichloroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only