missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-di-tert-butylphenol, 97%

Catalog No. p-7021045
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1321814 25 g
AAA1321822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1321814 Supplier Thermo Scientific Chemicals Supplier No. A1321814

CAS: 20834-61-1 | C14H21BrO | 285.225 g/mol

It is used in pharmaceutical intermediates and as medicine. When 2-Bromo-4,6-di-tert-butylphenol was treated with potassium ferricyanide in benzene 1,4-dihydro-4-bromo-2,4,6,8-tetra-tert-butyl-1-oxodibenzofuran was yielded with a small amount of 2,4,6,8-tetra-tert-butyldibenzofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical intermediates and as medicine. When 2-Bromo-4,6-di-tert-butylphenol was treated with potassium ferricyanide in benzene 1,4-dihydro-4-bromo-2,4,6,8-tetra-tert-butyl-1-oxodibenzofuran was yielded with a small amount of 2,4,6,8-tetra-tert-butyldibenzofuran.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 20834-61-1
Molecular Formula C14H21BrO
Molecular Weight (g/mol) 285.225
MDL Number MFCD00051597
InChI Key DIWZVAHZEOFSLS-UHFFFAOYSA-N
Synonym 2-bromo-4,6-di-tert-butylphenol, 2-bromo-4,6-di-t-butylphenol, 2,4-di-tert-butyl-6-bromophenol, phenol, 2-bromo-4,6-bis 1,1-dimethylethyl, phenol,2-bromo-4,6-bis 1,1-dimethylethyl, acmc-1crlp, 4,6-di-t-butyl-2-bromophenol, 6-bromo-2,4-di-t-butylphenol, 2-bromo4,6-di-tert-butylphenol, 2-bromo-4, 6-di-tert-butylphenol
PubChem CID 519822
IUPAC Name 2-bromo-4,6-ditert-butylphenol
SMILES CC(C)(C)C1=CC(=C(C(=C1)Br)O)C(C)(C)C

Specifications

Melting Point 55°C to 59°C
Quantity 25 g
Beilstein 2100150
Formula Weight 285.23
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,6-di-tert-butylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only