missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4,6-di-tert-butylphenol, 97%
Description
It is used in pharmaceutical intermediates and as medicine. When 2-Bromo-4,6-di-tert-butylphenol was treated with potassium ferricyanide in benzene 1,4-dihydro-4-bromo-2,4,6,8-tetra-tert-butyl-1-oxodibenzofuran was yielded with a small amount of 2,4,6,8-tetra-tert-butyldibenzofuran.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in pharmaceutical intermediates and as medicine. When 2-Bromo-4,6-di-tert-butylphenol was treated with potassium ferricyanide in benzene 1,4-dihydro-4-bromo-2,4,6,8-tetra-tert-butyl-1-oxodibenzofuran was yielded with a small amount of 2,4,6,8-tetra-tert-butyldibenzofuran.
Notes
Store at room temperature. Incompatible with oxidizing agents.
It is used in pharmaceutical intermediates and as medicine. When 2-Bromo-4,6-di-tert-butylphenol was treated with potassium ferricyanide in benzene 1,4-dihydro-4-bromo-2,4,6,8-tetra-tert-butyl-1-oxodibenzofuran was yielded with a small amount of 2,4,6,8-tetra-tert-butyldibenzofuran.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 55°C to 59°C |
| Quantity | 100 g |
| Beilstein | 2100150 |
| Formula Weight | 285.23 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Bromo-4,6-di-tert-butylphenol |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only