missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4,5-difluorotoluene 98.0+%, TCI America™
$63.38 - $199.46
Chemical Identifiers
| CAS | 875664-38-3 |
|---|---|
| Molecular Formula | C7H5BrF2 |
| Molecular Weight (g/mol) | 207.018 |
| MDL Number | MFCD07777160 |
| InChI Key | FKEURBCLFHOBDM-UHFFFAOYSA-N |
| Synonym | 2-bromo-4,5-difluorotoluene, 1-bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methylbromobenzene, ksc495c2l, bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methyl-1-bromobenzene, 1-brommo-4,5-difluoro-2-methyl-benzene, benzene,1-bromo-4,5-difluoro-2-methyl |
| PubChem CID | 44890785 |
| IUPAC Name | 1-bromo-4,5-difluoro-2-methylbenzene |
| SMILES | CC1=CC(=C(C=C1Br)F)F |
Chemical Identifiers
| 875664-38-3 | |
| 207.018 | |
| FKEURBCLFHOBDM-UHFFFAOYSA-N | |
| 44890785 | |
| CC1=CC(=C(C=C1Br)F)F |
| C7H5BrF2 | |
| MFCD07777160 | |
| 2-bromo-4,5-difluorotoluene, 1-bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methylbromobenzene, ksc495c2l, bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methyl-1-bromobenzene, 1-brommo-4,5-difluoro-2-methyl-benzene, benzene,1-bromo-4,5-difluoro-2-methyl | |
| 1-bromo-4,5-difluoro-2-methylbenzene |
Specifications
| 875664-38-3 | |
| C7H5BrF2 | |
| 1 g | |
| FKEURBCLFHOBDM-UHFFFAOYSA-N | |
| 1-bromo-4,5-difluoro-2-methylbenzene | |
| 44890785 | |
| ≥98.0% (GC) | |
| 2-Bromo-4,5-difluorotoluene |
| Yellow | |
| MFCD07777160 | |
| 2-bromo-4,5-difluorotoluene, 1-bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methylbromobenzene, ksc495c2l, bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methyl-1-bromobenzene, 1-brommo-4,5-difluoro-2-methyl-benzene, benzene,1-bromo-4,5-difluoro-2-methyl | |
| CC1=CC(=C(C=C1Br)F)F | |
| 207.018 | |
| 207.02 | |
| Liquid |
Safety and Handling
TSCA : No