CAS RN 875664-38-3
IUPAC Name:
1-bromo-4,5-difluoro-2-methylbenzene
Synonyms:
1-bromo-4,5-difluoro-2-methylbenzene
Molecular Weight (g/mol):
207.02
Molecular Formula:
C7H5BrF2
InChi Key:
FKEURBCLFHOBDM-UHFFFAOYSA-N
SMILES:
CC1=CC(F)=C(F)C=C1Br
1
–
1
of
1
results
1
2-Bromo-4,5-difluorotoluene 98.0+%, TCI America™
CAS: 875664-38-3 Molecular Formula: C7H5BrF2 Molecular Weight (g/mol): 207.018 MDL Number: MFCD07777160 InChI Key: FKEURBCLFHOBDM-UHFFFAOYSA-N Synonym: 2-bromo-4,5-difluorotoluene,1-bromo-4,5-difluoro-2-methyl-benzene,4,5-difluoro-2-methylbromobenzene,ksc495c2l,bromo-4,5-difluoro-2-methyl-benzene,4,5-difluoro-2-methyl-1-bromobenzene,1-brommo-4,5-difluoro-2-methyl-benzene,benzene,1-bromo-4,5-difluoro-2-methyl PubChem CID: 44890785 IUPAC Name: 1-bromo-4,5-difluoro-2-methylbenzene SMILES: CC1=CC(=C(C=C1Br)F)F