Learn More
2-Bromo-4,5-difluorophenylacetic acid, 98%
CAS: 883502-07-6 | C8H5BrF2O2 | 251.03 g/mol
$167.88 - $561.37
Chemical Identifiers
| CAS | 883502-07-6 |
|---|---|
| Molecular Formula | C8H5BrF2O2 |
| Molecular Weight (g/mol) | 251.03 |
| MDL Number | MFCD04973423 |
| InChI Key | FQSLXVRCDYUESO-UHFFFAOYSA-N |
| Synonym | 2-bromo-4,5-difluorophenylacetic acid, 2-2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenylaceticacid, pubchem2702, acmc-209qsj, 2-2-bromo-4,5-difluorophenyl aceticacid, benzeneacetic acid, 2-bromo-4,5-difluoro |
| PubChem CID | 3732964 |
| IUPAC Name | 2-(2-bromo-4,5-difluorophenyl)acetic acid |
| SMILES | OC(=O)CC1=CC(F)=C(F)C=C1Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6390203
|
Thermo Scientific Chemicals
H6390203 |
1 g |
N/A
|
|
|||||
|
AAH6390206
|
Thermo Scientific Chemicals
H6390206 |
5 g |
N/A
|
|
|||||
Description
2-Bromo-4,5-difluorophenylacetic acid is used as intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-4,5-difluorophenylacetic acid is used as intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 883502-07-6 | |
| 251.03 | |
| FQSLXVRCDYUESO-UHFFFAOYSA-N | |
| 3732964 | |
| OC(=O)CC1=CC(F)=C(F)C=C1Br |
| C8H5BrF2O2 | |
| MFCD04973423 | |
| 2-bromo-4,5-difluorophenylacetic acid, 2-2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenylaceticacid, pubchem2702, acmc-209qsj, 2-2-bromo-4,5-difluorophenyl aceticacid, benzeneacetic acid, 2-bromo-4,5-difluoro | |
| 2-(2-bromo-4,5-difluorophenyl)acetic acid |
Specifications
| 883502-07-6 | |
| C8H5BrF2O2 | |
| 1 g | |
| Slightly soluble in water. | |
| OC(=O)CC1=CC(F)=C(F)C=C1Br | |
| 251.03 | |
| 251.03 | |
| Solid |
| White | |
| MFCD04973423 | |
| 2-bromo-4,5-difluorophenylacetic acid, 2-2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenylaceticacid, pubchem2702, acmc-209qsj, 2-2-bromo-4,5-difluorophenyl aceticacid, benzeneacetic acid, 2-bromo-4,5-difluoro | |
| FQSLXVRCDYUESO-UHFFFAOYSA-N | |
| 2-(2-bromo-4,5-difluorophenyl)acetic acid | |
| 3732964 | |
| 98% | |
| 2-Bromo-4,5-difluorophenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only