CAS RN 883502-07-6
IUPAC Name:
2-(2-bromo-4,5-difluorophenyl)acetic acid
Synonyms:
(2-bromo-4,5-difluorophenyl)acetic acid
Molecular Weight (g/mol):
251.03
Molecular Formula:
C8H5BrF2O2
InChi Key:
FQSLXVRCDYUESO-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC(F)=C(F)C=C1Br
1
–
1
of
1
results
1
Promotions Available
2-Bromo-4,5-difluorophenylacetic acid, 98%
2-Bromo-4,5-difluorophenylacetic acid, 98%
CAS: 883502-07-6 Molecular Formula: C8H5BrF2O2 Molecular Weight (g/mol): 251.03 MDL Number: MFCD04973423 InChI Key: FQSLXVRCDYUESO-UHFFFAOYSA-N Synonym: 2-bromo-4,5-difluorophenylacetic acid,2-2-bromo-4,5-difluorophenyl acetic acid,2-bromo-4,5-difluorophenyl acetic acid,2-bromo-4,5-difluorophenylaceticacid,pubchem2702,acmc-209qsj,2-2-bromo-4,5-difluorophenyl aceticacid,benzeneacetic acid, 2-bromo-4,5-difluoro PubChem CID: 3732964 IUPAC Name: 2-(2-bromo-4,5-difluorophenyl)acetic acid SMILES: OC(=O)CC1=CC(F)=C(F)C=C1Br