missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluorophenol 98.0+%, TCI America™

Supplier:  TCI America B38455G

 View more versions of this product

Catalog No. B38455G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Bromo-4,5-difluorophenol
Yellow
MFCD00070751
2-bromo-4,5-diflouo phenol, 2-bromo-4,5-difluoro-phenol, phenol, 2-bromo-4,5-difluoro, pubchem2835, acmc-1bxcg, 2-bromo-4,5-difluoro, intermediates-zcf02247, 2-bromo-4,5-difluorophen, ksc494o4b, 2-bromo-4,5-difluorophenol
OC1=CC(F)=C(F)C=C1Br
208.99
208.99
Liquid
166281-37-4
C6H3BrF2O
5 g
FCYZOOHWUOEAOX-UHFFFAOYSA-N
2-bromo-4,5-difluorophenol
2736288
≥98.0% (GC)

Chemical Identifiers

166281-37-4
208.99
FCYZOOHWUOEAOX-UHFFFAOYSA-N
2736288
OC1=CC(F)=C(F)C=C1Br
C6H3BrF2O
MFCD00070751
2-bromo-4,5-diflouo phenol, 2-bromo-4,5-difluoro-phenol, phenol, 2-bromo-4,5-difluoro, pubchem2835, acmc-1bxcg, 2-bromo-4,5-difluoro, intermediates-zcf02247, 2-bromo-4,5-difluorophen, ksc494o4b, 2-bromo-4,5-difluorophenol
2-bromo-4,5-difluorophenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions