missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluorophenol 98.0+%, TCI America™

Catalog No. p-7123198
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38451G 1 g
B38455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38451G Supplier TCI America Supplier No. B38451G
Only null left

Chemical Identifiers

CAS 166281-37-4
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD00070751
InChI Key FCYZOOHWUOEAOX-UHFFFAOYSA-N
Synonym 2-bromo-4,5-diflouo phenol, 2-bromo-4,5-difluoro-phenol, phenol, 2-bromo-4,5-difluoro, pubchem2835, acmc-1bxcg, 2-bromo-4,5-difluoro, intermediates-zcf02247, 2-bromo-4,5-difluorophen, ksc494o4b, 2-bromo-4,5-difluorophenol
PubChem CID 2736288
IUPAC Name 2-bromo-4,5-difluorophenol
SMILES OC1=CC(F)=C(F)C=C1Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 208.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4,5-difluorophenol