missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluorobenzaldehyde, 98%

Catalog No. AAH6462103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6462103 1 g
AAH6462106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6462103 Supplier Thermo Scientific Chemicals Supplier No. H6462103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 476620-54-9 | C7H3BrF2O | 221.001 g/mol

It is an important intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 476620-54-9
Molecular Formula C7H3BrF2O
Molecular Weight (g/mol) 221.001
MDL Number MFCD16659662
InChI Key ZHSZEWFWVDLMDL-UHFFFAOYSA-N
Synonym benzaldehyde, 2-bromo-4,5-difluoro, acmc-209ka1, 2-bromo-4,5-difluoro-benzaldehyde
PubChem CID 21867425
IUPAC Name 2-bromo-4,5-difluorobenzaldehyde
SMILES C1=C(C(=CC(=C1F)F)Br)C=O

Specifications

Color White
Quantity 1 g
Beilstein 20365202
Solubility Information Slightly soluble in water.
Formula Weight 221
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Bromo-4,5-difluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only