accessibility menu, dialog, popup

UserName

CAS RN 476620-54-9

IUPAC Name: 2-bromo-4,5-difluorobenzaldehyde
Synonyms: 2-bromo-4,5-difluorobenzaldehyde
Molecular Weight (g/mol): 221
Molecular Formula: C7H3BrF2O
InChi Key: ZHSZEWFWVDLMDL-UHFFFAOYSA-N
SMILES: FC1=C(F)C=C(C=O)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results