missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals

Catalog No. p-7055670
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6430903 1 g
AAH6430906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6430903 Supplier Thermo Scientific Chemicals Supplier No. H6430903

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58458-10-9
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.023
MDL Number MFCD01656033
InChI Key XIUJCAJFRWDEKE-UHFFFAOYSA-N
Synonym 2-bromo-3-trifluoromethyl aniline, 3-amino-2-bromobenzotrifluoride, 2-bromo-3-trifluoromethyl benzenamine, 2-bromo-3-trifluoromethyl phenylamine, benzenamine, 2-bromo-3-trifluoromethyl, pubchem4829, ksc275a9t, 2-bromo-3-trifluoromethyl-aniline, 3-amino-2-bromobenzotrifluoride 2-bromo-3-trifluoromethyl aniline
PubChem CID 94058
IUPAC Name 2-bromo-3-(trifluoromethyl)aniline
SMILES C1=CC(=C(C(=C1)N)Br)C(F)(F)F

Specifications

Color White
Quantity 1 g
UN Number UN2811
Beilstein 2835356
Formula Weight 240.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-3-(trifluoromethyl)aniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 261-261-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only