CAS RN 58458-10-9
IUPAC Name:
2-bromo-3-(trifluoromethyl)aniline
Synonyms:
2-bromo-3-(trifluoromethyl)aniline
Molecular Weight (g/mol):
240.02
Molecular Formula:
C7H5BrF3N
InChi Key:
XIUJCAJFRWDEKE-UHFFFAOYSA-N
SMILES:
NC1=CC=CC(=C1Br)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
2-Bromo-3-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals
2-Bromo-3-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals
CAS: 58458-10-9 Molecular Formula: C7H5BrF3N Molecular Weight (g/mol): 240.023 MDL Number: MFCD01656033 InChI Key: XIUJCAJFRWDEKE-UHFFFAOYSA-N Synonym: 2-bromo-3-trifluoromethyl aniline,3-amino-2-bromobenzotrifluoride,2-bromo-3-trifluoromethyl benzenamine,2-bromo-3-trifluoromethyl phenylamine,benzenamine, 2-bromo-3-trifluoromethyl,pubchem4829,ksc275a9t,2-bromo-3-trifluoromethyl-aniline,3-amino-2-bromobenzotrifluoride 2-bromo-3-trifluoromethyl aniline PubChem CID: 94058 IUPAC Name: 2-bromo-3-(trifluoromethyl)aniline SMILES: C1=CC(=C(C(=C1)N)Br)C(F)(F)F