missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7122873
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33225G 5 g
B332225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33225G Supplier TCI America Supplier No. B33225G

Chemical Identifiers

CAS 573-54-6
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD00074899
InChI Key WTDJEGSXLFHZPY-UHFFFAOYSA-N
Synonym 2-bromo-3-nitrobenzoic acid, benzoic acid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoicacid, 2-bromo-3-nitro-benzoic acid, pubchem4714, acmc-1an8y, ksc273m5t, benzoicacid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoic acid, technical grade
PubChem CID 68452
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])Br)C(=O)O

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 5 g
Formula Weight 246.02
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-nitrobenzoic Acid