CAS RN 573-54-6
IUPAC Name:
2-bromo-3-nitrobenzoic acid
Synonyms:
2-bromo-3-nitrobenzoic acid
Molecular Weight (g/mol):
246.02
Molecular Formula:
C7H4BrNO4
InChi Key:
WTDJEGSXLFHZPY-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC(=C1Br)[N+]([O-])=O
1
–
1
of
1
results
1
2-Bromo-3-nitrobenzoic Acid 98.0+%, TCI America™
CAS: 573-54-6 Molecular Formula: C7H4BrNO4 Molecular Weight (g/mol): 246.016 MDL Number: MFCD00074899 InChI Key: WTDJEGSXLFHZPY-UHFFFAOYSA-N Synonym: 2-bromo-3-nitrobenzoic acid,benzoic acid, 2-bromo-3-nitro,2-bromo-3-nitrobenzoicacid,2-bromo-3-nitro-benzoic acid,pubchem4714,acmc-1an8y,ksc273m5t,benzoicacid, 2-bromo-3-nitro,2-bromo-3-nitrobenzoic acid, technical grade PubChem CID: 68452 SMILES: C1=CC(=C(C(=C1)[N+](=O)[O-])Br)C(=O)O