missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3'-nitroacetophenone 97.0+%, TCI America™
Supplier: TCI America B285525G
Specifications
| 2-Bromo-3′-nitroacetophenone | |
| 94°C | |
| MFCD00024512 | |
| 3261 | |
| GZHPNIQBPGUSSX-UHFFFAOYSA-N | |
| 2-bromo-1-(3-nitrophenyl)ethanone | |
| 75213 | |
| ≥97.0% (GC,T) |
| 2227-64-7 | |
| C8H6BrNO3 | |
| 25 g | |
| 2-bromo-1-3-nitrophenyl ethanone, 2-bromo-3'-nitroacetophenone, 3-nitrophenacyl bromide, 3-nitrophenacylbromide, 2-bromo-1-3-nitrophenyl ethan-1-one, a-bromo-3-nitroacetophenone, 2-bromo-3-nitroacetophenone, alpha-bromo-3'-nitroacetophenone, acetophenone, 2-bromo-3'-nitro, m-nitrophenacyl bromide | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)CBr | |
| 244.044 | |
| 244.04 | |
| Crystalline Powder |
Chemical Identifiers
| 2227-64-7 | |
| 244.044 | |
| GZHPNIQBPGUSSX-UHFFFAOYSA-N | |
| 75213 | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)CBr |
| C8H6BrNO3 | |
| MFCD00024512 | |
| 2-bromo-1-3-nitrophenyl ethanone, 2-bromo-3'-nitroacetophenone, 3-nitrophenacyl bromide, 3-nitrophenacylbromide, 2-bromo-1-3-nitrophenyl ethan-1-one, a-bromo-3-nitroacetophenone, 2-bromo-3-nitroacetophenone, alpha-bromo-3'-nitroacetophenone, acetophenone, 2-bromo-3'-nitro, m-nitrophenacyl bromide | |
| 2-bromo-1-(3-nitrophenyl)ethanone |
Safety and Handling
EINECSNumber : (3)-1248
TSCA : No