accessibility menu, dialog, popup

UserName

ONOOBr

CAS RN 2227-64-7

IUPAC Name: 2-bromo-1-(3-nitrophenyl)ethan-1-one
Synonyms: 2-bromo-1-(3-nitrophenyl)ethanone
Molecular Weight (g/mol): 244.04
Molecular Formula: C8H6BrNO3
InChi Key: GZHPNIQBPGUSSX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)C(=O)CBr
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)