missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-nitroacetophenone 97.0+%, TCI America™

Catalog No. p-7119808
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B28555G 5 g
B285525G 25 g
2 options

Catalog No. B28555G

Supplier: TCI America B28555G

Only null left

Chemical Identifiers

CAS 2227-64-7
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.044
MDL Number MFCD00024512
InChI Key GZHPNIQBPGUSSX-UHFFFAOYSA-N
Synonym 2-bromo-1-3-nitrophenyl ethanone, 2-bromo-3'-nitroacetophenone, 3-nitrophenacyl bromide, 3-nitrophenacylbromide, 2-bromo-1-3-nitrophenyl ethan-1-one, a-bromo-3-nitroacetophenone, 2-bromo-3-nitroacetophenone, alpha-bromo-3'-nitroacetophenone, acetophenone, 2-bromo-3'-nitro, m-nitrophenacyl bromide
PubChem CID 75213
IUPAC Name 2-bromo-1-(3-nitrophenyl)ethanone
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)CBr

Specifications

Melting Point 94°C
Quantity 5 g
UN Number 3261
Formula Weight 244.04
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3′-nitroacetophenone
EINECSNumber (3)-1248
TSCA No