missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-hydroxypyridine 98.0+%, TCI America™
Supplier: TCI America B147825G
Specifications
| 2-Bromo-3-hydroxypyridine | |
| 181°C | |
| MFCD00006220 | |
| 2-bromo-3-hydroxypyridine, 2-bromo-3-pyridinol, 3-pyridinol, 2-bromo, 2-bromo-pyridin-3-ol, 3-hydroxy-2-bromopyridine, unii-9dab85h359, 2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo3-pyridinol | |
| OC1=CC=CN=C1Br | |
| 174.00 | |
| 174.00 | |
| Crystalline Powder |
| 6602-32-0 | |
| C5H4BrNO | |
| 25 g | |
| YKHQFTANTNMYPP-UHFFFAOYSA-N | |
| 2-bromopyridin-3-ol | |
| 23070 | |
| ≥98.0% (T) |
Chemical Identifiers
| 6602-32-0 | |
| 174.00 | |
| YKHQFTANTNMYPP-UHFFFAOYSA-N | |
| 23070 | |
| OC1=CC=CN=C1Br |
| C5H4BrNO | |
| MFCD00006220 | |
| 2-bromo-3-hydroxypyridine, 2-bromo-3-pyridinol, 3-pyridinol, 2-bromo, 2-bromo-pyridin-3-ol, 3-hydroxy-2-bromopyridine, unii-9dab85h359, 2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo3-pyridinol | |
| 2-bromopyridin-3-ol |
Safety and Handling
RTECSNumber : UU7705000
TSCA : No