accessibility menu, dialog, popup

UserName

OHNBr

CAS RN 6602-32-0

IUPAC Name: 2-bromopyridin-3-ol
Synonyms: 3-pyridinol, 2-bromo-
Molecular Weight (g/mol): 174
Molecular Formula: C5H4BrNO
InChi Key: YKHQFTANTNMYPP-UHFFFAOYSA-N
SMILES: OC1=CC=CN=C1Br
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (4)