missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-hydroxypyridine 98.0+%, TCI America™

Catalog No. p-7119075
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14785G 5 g
B147825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14785G Supplier TCI America Supplier No. B14785G
Only null left

Chemical Identifiers

CAS 6602-32-0
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 174.00
MDL Number MFCD00006220
InChI Key YKHQFTANTNMYPP-UHFFFAOYSA-N
Synonym 2-bromo-3-hydroxypyridine, 2-bromo-3-pyridinol, 3-pyridinol, 2-bromo, 2-bromo-pyridin-3-ol, 3-hydroxy-2-bromopyridine, unii-9dab85h359, 2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo3-pyridinol
PubChem CID 23070
IUPAC Name 2-bromopyridin-3-ol
SMILES OC1=CC=CN=C1Br

Specifications

Melting Point 181°C
Quantity 5 g
Formula Weight 174.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-hydroxypyridine
RTECSNumber UU7705000
TSCA No