missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluorobenzyl bromide, 98%

Catalog No. p-7041670
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6363203 1 g
AAH6363206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6363203 Supplier Thermo Scientific Chemicals Supplier No. H6363203

CAS: 1184918-22-6 | C7H5Br2F | 267.92 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1184918-22-6
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.92
MDL Number MFCD12911576
InChI Key ZJSRIMJDFLFPJI-UHFFFAOYSA-N
Synonym 2-bromo-1-bromomethyl-3-fluorobenzene, 2-bromo-3-fluorobenzyl bromide, 2-bromo-3-bromomethyl-1-fluorobenzene, 2-bromo-3-fluorobenzylbromide
PubChem CID 44181799
IUPAC Name 2-bromo-1-(bromomethyl)-3-fluorobenzene
SMILES FC1=CC=CC(CBr)=C1Br

Specifications

Color White
Quantity 1 g
UN Number UN3261
Solubility Information Slightly soluble in water.
Formula Weight 267.92
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-3-fluorobenzyl bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only