CAS RN 1184918-22-6
IUPAC Name:
2-bromo-1-(bromomethyl)-3-fluorobenzene
Synonyms:
2-bromo-1-(bromomethyl)-3-fluorobenzene
Molecular Weight (g/mol):
267.92
Molecular Formula:
C7H5Br2F
InChi Key:
ZJSRIMJDFLFPJI-UHFFFAOYSA-N
SMILES:
FC1=CC=CC(CBr)=C1Br
1
–
1
of
1
results
1
Promotions Available
2-Bromo-3-fluorobenzyl bromide, 98%
2-Bromo-3-fluorobenzyl bromide, 98%
CAS: 1184918-22-6 Molecular Formula: C7H5Br2F Molecular Weight (g/mol): 267.92 MDL Number: MFCD12911576 InChI Key: ZJSRIMJDFLFPJI-UHFFFAOYSA-N Synonym: 2-bromo-1-bromomethyl-3-fluorobenzene,2-bromo-3-fluorobenzyl bromide,2-bromo-3-bromomethyl-1-fluorobenzene,2-bromo-3-fluorobenzylbromide PubChem CID: 44181799 IUPAC Name: 2-bromo-1-(bromomethyl)-3-fluorobenzene SMILES: FC1=CC=CC(CBr)=C1Br