missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluorobenzaldehyde, 98%

Catalog No. p-7055745
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6440006 5 g
AAH6440014 25 g
AAH6440022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6440006 Supplier Thermo Scientific Chemicals Supplier No. H6440006

CAS: 891180-59-9 | C7H4BrFO | 203.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 891180-59-9
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD07782044
InChI Key CYJBMGYWRHGZBR-UHFFFAOYSA-N
Synonym 2-bromo-3-fluoro-benzaldehyde, benzaldehyde, 2-bromo-3-fluoro, pubchem1417, pubchem22500, acmc-1bwut, ksc494e8j, 6-fluoro-2-formylbromobenzene
PubChem CID 24820504
IUPAC Name 2-bromo-3-fluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)F)Br)C=O

Specifications

Quantity 5 g
Beilstein 10529128
Sensitivity Light sensitive
Formula Weight 203.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-3-fluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only