missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-butene (stabilized with Copper chip) 98.0+%, TCI America™

Supplier:  TCI America B141225G

 View more versions of this product

Catalog No. B141225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Bromo-2-butene (stabilized with Copper chip)
C4H7Br
25 g
2-bromo-2-butene, trans-2-bromo-2-butene, 2-butene, 2-bromo, e-2-bromobut-2-ene, 2-bromo-2-butene cis, e-2-bromo-2-butene, 1-methyl-1-propenyl bromide, cis-2-bromo-2-butene, e-2-bromo-but-2-ene, 2e-2-bromo-2-butene #
C\C=C(/C)Br
135.00
135.00
Liquid
13294-71-8
MFCD00000141
1993
UILZQFGKPHAAOU-ONEGZZNKSA-N
(2E)-2-bromobut-2-ene
5364387
≥98.0% (GC)

Chemical Identifiers

13294-71-8
135.00
UILZQFGKPHAAOU-ONEGZZNKSA-N
5364387
C\C=C(/C)Br
C4H7Br
MFCD00000141
2-bromo-2-butene, trans-2-bromo-2-butene, 2-butene, 2-bromo, e-2-bromobut-2-ene, 2-bromo-2-butene cis, e-2-bromo-2-butene, 1-methyl-1-propenyl bromide, cis-2-bromo-2-butene, e-2-bromo-but-2-ene, 2e-2-bromo-2-butene #
(2E)-2-bromobut-2-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions