missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-butene (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. p-7119040
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B141225G 25 g
B1412500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B141225G Supplier TCI America Supplier No. B141225G
Only null left

Chemical Identifiers

CAS 13294-71-8
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.00
MDL Number MFCD00000141
InChI Key UILZQFGKPHAAOU-ONEGZZNKSA-N
Synonym 2-bromo-2-butene, trans-2-bromo-2-butene, 2-butene, 2-bromo, e-2-bromobut-2-ene, 2-bromo-2-butene cis, e-2-bromo-2-butene, 1-methyl-1-propenyl bromide, cis-2-bromo-2-butene, e-2-bromo-but-2-ene, 2e-2-bromo-2-butene #
PubChem CID 5364387
IUPAC Name (2E)-2-bromobut-2-ene
SMILES C\C=C(/C)Br

Specifications

Quantity 25 g
UN Number 1993
Formula Weight 135.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-2-butene (stabilized with Copper chip)