missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-tetralone 97.0+%, TCI America™

Catalog No. p-7124162
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56981G 1 g
B56985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56981G Supplier TCI America Supplier No. B56981G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13672-07-6
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.085
MDL Number MFCD00089999
InChI Key AYNCWMIFKFADCZ-UHFFFAOYSA-N
Synonym 2-Bromo-3,4-dihydro-1(2H)-naphthalenone
PubChem CID 101120
IUPAC Name 2-bromo-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=CC=CC=C2C(=O)C1Br

Specifications

Melting Point 39°C
Quantity 1 g
Formula Weight 225.09
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1-tetralone