accessibility menu, dialog, popup

UserName

BrO

CAS RN 13672-07-6

IUPAC Name: 2-bromo-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 2-bromo-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 225.09
Molecular Formula: C10H9BrO
InChi Key: AYNCWMIFKFADCZ-UHFFFAOYNA-N
SMILES: BrC1CCC2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results