missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-chloro-4-fluorobenzene 98.0+%, TCI America™

Catalog No. p-7119915
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30485G 5 g
B304825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30485G Supplier TCI America Supplier No. B30485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 201849-15-2
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.44
MDL Number MFCD00061120
InChI Key FOCCSIJMXBTKHD-UHFFFAOYSA-N
Synonym 2-chloro-5-fluorobromobenzene, 1-bromo-2-chloro-5-fluorobenzene, 3-bromo-4-chlorofluorobenzene, 2-bromo-1-chloro-4-fluoro-benzene, benzene, 2-bromo-1-chloro-4-fluoro, pubchem4270, acmc-209f6i, ksc494o8h, 1-chloro-2-bromo-4-fluorobenzene, 2-bromo-4-fluorochlorobenzene
PubChem CID 2773263
IUPAC Name 2-bromo-1-chloro-4-fluorobenzene
SMILES FC1=CC(Br)=C(Cl)C=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 209.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-1-chloro-4-fluorobenzene