accessibility menu, dialog, popup

UserName

CAS RN 201849-15-2

IUPAC Name: 2-bromo-1-chloro-4-fluorobenzene
Synonyms: 2-bromo-1-chloro-4-fluorobenzene
Molecular Weight (g/mol): 209.44
Molecular Formula: C6H3BrClF
InChi Key: FOCCSIJMXBTKHD-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(Cl)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results