missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,4-difluorobenzene, 98%

Catalog No. p-7020863
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1296306 5 g
AAA1296314 25 g
AAA1296322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1296306 Supplier Thermo Scientific Chemicals Supplier No. A1296306
Only null left

CAS: 399-94-0 | C6H3BrF2 | 192.991 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 399-94-0
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.991
MDL Number MFCD00000345
InChI Key XCRCSPKQEDMVBO-UHFFFAOYSA-N
Synonym 1-bromo-2,5-difluorobenzene, 2,5-difluorobromobenzene, benzene, 2-bromo-1,4-difluoro, 1,4-difluoro-2-bromobenzene, 2-bromo-1,4-difluoro-benzene, fr df be, pubchem2302, acmc-1ad5i, 1-bromo-2,5 difluorobenzene, 2,5-difluoro-1-bromobenzene
PubChem CID 67862
IUPAC Name 2-bromo-1,4-difluorobenzene
SMILES C1=CC(=C(C=C1F)Br)F

Specifications

Melting Point -31°C
Density 1.714
Boiling Point 73°C to 75°C (65 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.508
Quantity 5 g
Beilstein 1680893
Formula Weight 192.99
Percent Purity 98%
Chemical Name or Material 2-Bromo-1,4-difluorobenzene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-920-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only