missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3-diethylbenzene, 94%

Catalog No. AAL0348206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0348206 5 g
AAL0348203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0348206 Supplier Thermo Scientific Chemicals Supplier No. L0348206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65232-57-7 | C10H13Br | 213.12 g/mol

2-Bromo-1,3-diethylbenzene is used as raw material in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1,3-diethylbenzene is used as raw material in synthetic organic chemistry.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 65232-57-7
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.12
MDL Number MFCD00017770
InChI Key FPWGIABDOFXETH-UHFFFAOYSA-N
Synonym 2,6-diethylbromobenzene, 1-bromo-2,6-diethylbenzene, 2,6-diethyl bromobenzene, benzene, 2-bromo-1,3-diethyl, acmc-20ao3d, 2,6-diethylphenylbromide, 2,6-diethylphenyl bromide, 1,3-diethyl-2-bromobenzene, 2-bromo-1,3-diethyl-benzene, #
PubChem CID 522160
IUPAC Name 2-bromo-1,3-diethylbenzene
SMILES CCC1=CC=CC(CC)=C1Br

Specifications

Density 1.264
Boiling Point 78°C to 80°C (2 mmHg)
Refractive Index 1.544
Quantity 5 g
Beilstein 2501180
Solubility Information Insoluble in water.
Formula Weight 213.12
Percent Purity 94%
Chemical Name or Material 2-Bromo-1,3-diethylbenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only