accessibility menu, dialog, popup

UserName

CAS RN 65232-57-7

IUPAC Name: 2-bromo-1,3-diethylbenzene
Synonyms: 2-bromo-1,3-diethylbenzene
Molecular Weight (g/mol): 213.12
Molecular Formula: C10H13Br
InChi Key: FPWGIABDOFXETH-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(CC)=C1Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results