missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene, 97%

Catalog No. AAH3258003
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3258003 1 g
AAH3258009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3258003 Supplier Thermo Scientific Chemicals Supplier No. H3258003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 118754-55-5 | C7H2BrCl2F3O | 309.891 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118754-55-5
Molecular Formula C7H2BrCl2F3O
Molecular Weight (g/mol) 309.891
MDL Number MFCD03412210
InChI Key CAAJZKZQEJSRJO-UHFFFAOYSA-N
Synonym 2-bromo-1,3-dichloro-5-trifluoromethoxy benzene, 2,6-dichloro-4-trifluoromethoxy bromobenzene, 4-bromo-3,5-dichloro-1-trifluoromethoxy benzene, benzene,2-bromo-1,3-dichloro-5-trifluoromethoxy, acmc-1c5si
PubChem CID 18947696
IUPAC Name 2-bromo-1,3-dichloro-5-(trifluoromethoxy)benzene
SMILES C1=C(C=C(C(=C1Cl)Br)Cl)OC(F)(F)F

Specifications

Quantity 1 g
Formula Weight 309.9
Percent Purity 97%
Chemical Name or Material 2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only