accessibility menu, dialog, popup

UserName

CAS RN 118754-55-5

IUPAC Name: 2-bromo-1,3-dichloro-5-(trifluoromethoxy)benzene
Synonyms: 2-bromo-1,3-dichloro-5-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 309.89
Molecular Formula: C7H2BrCl2F3O
InChi Key: CAAJZKZQEJSRJO-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC(Cl)=C(Br)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results