missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3-bis(bromomethyl)benzene 98.0+%, TCI America™

Catalog No. p-7123240
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39135G 5 g
B391325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39135G Supplier TCI America Supplier No. B39135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25006-88-6
Molecular Formula C8H7Br3
Molecular Weight (g/mol) 342.86
MDL Number MFCD00229246
InChI Key HSYMWXREIPJGHX-UHFFFAOYSA-N
PubChem CID 621912
IUPAC Name 2-bromo-1,3-bis(bromomethyl)benzene
SMILES BrCC1=CC=CC(CBr)=C1Br

Specifications

Melting Point 106°C
Color White
Quantity 5 g
UN Number 3261
Synonym alpha,alpha′C,2-Tribromo-m-xylene
Formula Weight 342.86
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1,3-bis(bromomethyl)benzene