accessibility menu, dialog, popup

UserName

CAS RN 25006-88-6

IUPAC Name: 2-bromo-1,3-bis(bromomethyl)benzene
Synonyms: 2-bromo-1,3-bis(bromomethyl)benzene
Molecular Weight (g/mol): 342.86
Molecular Formula: C8H7Br3
InChi Key: HSYMWXREIPJGHX-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC(CBr)=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results