missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3,5-trifluorobenzene, 98+%

Catalog No. AAA1992006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1992006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1992006 Supplier Thermo Scientific Chemicals Supplier No. A1992006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2367-76-2 | C6H2BrF3 | 210.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 2367-76-2
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.98
MDL Number MFCD00000334
InChI Key PZBSPSOGEVCRQI-UHFFFAOYSA-N
Synonym 1-bromo-2,4,6-trifluorobenzene, 2,4,6-trifluorobromobenzene, 2-bromo-1,3,5-trifluoro-benzene, benzene, 2-bromo-1,3,5-trifluoro, 1-bromo-2,4,6-trifluoro benzene, 1,3,5-trifluoro-2-bromobenzene, fr cf ef be, pubchem4306, acmc-1cbzk, intermediates-zcf02629
PubChem CID 75398
IUPAC Name 2-bromo-1,3,5-trifluorobenzene
SMILES FC1=CC(F)=C(Br)C(F)=C1

Specifications

Melting Point 3°C to 4°C
Density 1.79
Boiling Point 140°C to 141°C
Flash Point 58°C (136°F)
Refractive Index 1.485
Quantity 5 g
UN Number UN1993
Beilstein 2502668
Formula Weight 210.98
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-1,3,5-trifluorobenzene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 219-125-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only