accessibility menu, dialog, popup

UserName

FFBrF

CAS RN 2367-76-2

IUPAC Name: 2-bromo-1,3,5-trifluorobenzene
Synonyms: 2-bromo-1,3,5-trifluorobenzene
Molecular Weight (g/mol): 210.98
Molecular Formula: C6H2BrF3
InChi Key: PZBSPSOGEVCRQI-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(Br)C(F)=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)