missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[Bis(4-hydroxyphenyl)methyl]benzyl Alcohol 98.0+%, TCI America™
$262.83 - $915.25
Chemical Identifiers
| CAS | 81-92-5 |
|---|---|
| Molecular Formula | C20H18O3 |
| Molecular Weight (g/mol) | 306.36 |
| MDL Number | MFCD00059144 |
| InChI Key | CREICILGVGNQBH-UHFFFAOYSA-N |
| Synonym | Phenolphthalol |
| PubChem CID | 66495 |
| IUPAC Name | 4-{[2-(hydroxymethyl)phenyl](4-hydroxyphenyl)methyl}phenol |
| SMILES | OCC1=CC=CC=C1C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1 |
Chemical Identifiers
| 81-92-5 | |
| 306.36 | |
| CREICILGVGNQBH-UHFFFAOYSA-N | |
| 66495 | |
| OCC1=CC=CC=C1C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1 |
| C20H18O3 | |
| MFCD00059144 | |
| Phenolphthalol | |
| 4-{[2-(hydroxymethyl)phenyl](4-hydroxyphenyl)methyl}phenol |
Specifications
| 81-92-5 | |
| White | |
| MFCD00059144 | |
| Phenolphthalol | |
| OCC1=CC=CC=C1C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1 | |
| 306.36 | |
| 306.36 | |
| Crystalline Powder |
| 204°C | |
| C20H18O3 | |
| 5 g | |
| CREICILGVGNQBH-UHFFFAOYSA-N | |
| 4-{[2-(hydroxymethyl)phenyl](4-hydroxyphenyl)methyl}phenol | |
| 66495 | |
| ≥98.0% (GC) | |
| 2-[Bis(4-hydroxyphenyl)methyl]benzyl Alcohol |
Safety and Handling
RTECSNumber : DA4912000
TSCA : No