CAS RN 81-92-5
IUPAC Name:
4-{[2-(hydroxymethyl)phenyl](4-hydroxyphenyl)methyl}phenol
Synonyms:
phenolphthalol
Molecular Weight (g/mol):
306.36
Molecular Formula:
C20H18O3
InChi Key:
CREICILGVGNQBH-UHFFFAOYSA-N
SMILES:
OCC1=CC=CC=C1C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
2-[Bis(4-hydroxyphenyl)methyl]benzyl Alcohol 98.0+%, TCI America™
CAS: 81-92-5 Molecular Formula: C20H18O3 Molecular Weight (g/mol): 306.36 MDL Number: MFCD00059144 InChI Key: CREICILGVGNQBH-UHFFFAOYSA-N Synonym: Phenolphthalol PubChem CID: 66495 IUPAC Name: 4-{[2-(hydroxymethyl)phenyl](4-hydroxyphenyl)methyl}phenol SMILES: OCC1=CC=CC=C1C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1