missing translation for 'onlineSavingsMsg'
Learn More

2-[Bis(4-hydroxyphenyl)methyl]benzyl Alcohol 98.0+%, TCI America™

Catalog No. B01005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B01005G 5 g
B010025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B01005G Supplier TCI America Supplier No. B01005G
Only null left

Chemical Identifiers

CAS 81-92-5
Molecular Formula C20H18O3
Molecular Weight (g/mol) 306.36
MDL Number MFCD00059144
InChI Key CREICILGVGNQBH-UHFFFAOYSA-N
Synonym Phenolphthalol
PubChem CID 66495
IUPAC Name 4-{[2-(hydroxymethyl)phenyl](4-hydroxyphenyl)methyl}phenol
SMILES OCC1=CC=CC=C1C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 204°C
Color White
Quantity 5 g
Formula Weight 306.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-[Bis(4-hydroxyphenyl)methyl]benzyl Alcohol
RTECSNumber DA4912000
TSCA No