missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzylthio)ethanol, 98%

Catalog No. AAL1227414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1227414 25 g
AAL1227406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1227414 Supplier Thermo Scientific Chemicals Supplier No. L1227414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26524-88-9 | C9H12OS | 168.254 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26524-88-9
Molecular Formula C9H12OS
Molecular Weight (g/mol) 168.254
MDL Number MFCD00014039
InChI Key ANMHSIXPUAKNLM-UHFFFAOYSA-N
Synonym 2-benzylthio ethanol, 2-hydroxyethyl benzyl sulfide, 2-benzylsulfanyl ethanol, 2-hydroxyethyl-benzyl sulphide, 2-benzylsulfanyl ethan-1-ol, 2-phenylmethylthio ethanol, 2-phenylmethylsulfanyl ethanol, 2-phenylmethylthio ethan-1-ol, 2-hydroxyethylbenzylsulfide, 2-benzylthioethanol
PubChem CID 77486
IUPAC Name 2-benzylsulfanylethanol
SMILES C1=CC=C(C=C1)CSCCO

Specifications

Boiling Point 168°C to 169°C (18 mmHg)
Refractive Index 1.576
Linear Formula C6H5CH2S(CH2)2OH
Quantity 25 g
Beilstein 2356895
Formula Weight 168.25
Percent Purity 98%
Chemical Name or Material 2-(Benzylthio)ethanol
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only